Invitrogen
Manufacturer Code:PA5104026
Catalog # PIPA5104026
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KCNG4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4921535I01Rik AW049024 KCNG3 Kcng4 KV6.3 KV6.4 potassium channel voltage gated modifier subfamily G member 4 potassium voltage-gated channel modifier subfamily G member 4 potassium voltage-gated channel subfamily G member 4 potassium voltage-gated channel subfamily G member 4 voltage-gated potassium channel Kv6.3 voltage-gated potassium channel subunit Kv6.3 voltage-gated potassium channel subunit Kv6.4; potassium channel, voltage gated modifier subfamily G, member 4; Potassium voltage-gated channel subfamily G member 4; potassium voltage-gated channel, subfamily G, member 4; voltage-gated potassium channel Kv6.3; voltage-gated potassium channel subunit Kv6.3; Voltage-gated potassium channel subunit Kv6.4
Gene Aliases: 4921535I01Rik; AW049024; KCNG3; KCNG4; KV6.3; KV6.4
UniProt ID: (Human) Q8TDN1, (Mouse) Q80XM3
Entrez Gene ID: (Human) 93107, (Rat) 307900, (Mouse) 66733
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.