Invitrogen
Manufacturer Code:PA559033
Catalog # PIPA559033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KCNG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K13 KCNG KV6.1 kH2 potassium channel KH2 potassium channel Kv6.1 potassium voltage-gated channel subfamily G member 1 voltage-gated potassium channel subunit Kv6.1; kH2; potassium channel KH2; potassium channel Kv6.1; potassium channel, voltage gated modifier subfamily G, member 1; Potassium voltage-gated channel subfamily G member 1; potassium voltage-gated channel, subfamily G, member 1; Voltage-gated potassium channel subunit Kv6.1
Gene Aliases: K13; KCNG; KCNG1; kH2; KV6.1
UniProt ID: (Human) A8K3S4
Entrez Gene ID: (Human) 3755
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.