Abnova
Manufacturer Code:PAB23612
Catalog # 89-125-319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human KCNG1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant KCNG1.
For Research Use Only
Protein Aliases: K13; KCNG; kH2; KV6.1; MGC12878; potassium channel KH2; potassium channel Kv6.1; potassium channel, voltage gated modifier subfamily G, member 1; Potassium voltage-gated channel subfamily G member 1; potassium voltage-gated channel, subfamily G, member 1; Voltage-gated potassium channel subunit Kv6.1
Gene Aliases: K13; KCNG; KCNG1; kH2; KV6.1
UniProt ID: (Human) A8K3S4
Entrez Gene ID: (Human) 3755
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.