Invitrogen
Manufacturer Code:PA584543
Catalog # PIPA584543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KCNF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IK8 KCNF kH1 Kv5.1; kH1; potassium channel KH1; Potassium voltage-gated channel subfamily F member 1; Voltage-gated potassium channel subunit Kv5.1
Gene Aliases: IK8; KCNF; KCNF1; kH1; KV5.1
UniProt ID: (Human) Q9H3M0
Entrez Gene ID: (Human) 3754
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.