Invitrogen
Manufacturer Code:PA5104025
Catalog # PIPA5104025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KCNE3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2210017H05Rik cardiac voltage-gated potassium channel accessory subunit HOKPP HYPP KCNE3 minimum potassium ion channel-related peptide 2 MinK-related peptide 2 MiRP2 Potassium channel subunit beta MiRP2 potassium channel voltage gated subfamily E regulatory beta subunit 3 potassium channel voltage-gated Isk-related subfamily E regulatory beta subunit 3 potassium voltage-gated channel subfamily E member 3 potassium voltage-gated channel subfamily E regulatory subunit 3 potassium voltage-gated channel Isk-related family member 3 potassium voltage-gated channel Isk-related subfamily gene 3 potassium voltage-gated channel Isk-related subfamily member 3 voltage-gated K+ channel subunit MIRP2; cardiac voltage-gated potassium channel accessory subunit; Minimum potassium ion channel-related peptide 2; MinK-related peptide 2; Potassium channel subunit beta MiRP2; potassium channel, voltage gated subfamily E regulatory beta subunit 3; Potassium voltage-gated channel subfamily E member 3; potassium voltage-gated channel, Isk-related family, member 3; potassium voltage-gated channel, Isk-related subfamily, gene 3; potassium voltage-gated channel, Isk-related subfamily, member 3; voltage-gated K+ channel subunit MIRP2
Gene Aliases: 2210017H05Rik; HOKPP; HYPP; KCNE3; MiRP2
UniProt ID: (Human) Q9Y6H6, (Rat) Q9JJV7, (Mouse) Q9WTW2
Entrez Gene ID: (Human) 10008, (Rat) 63883, (Mouse) 57442
Molecular Function:
ion channel
potassium channel
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.