Abnova
Manufacturer Code:PAB20775
Catalog # 89-122-276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human KCNE3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant KCNE3.
For Research Use Only
Protein Aliases: cardiac voltage-gated potassium channel accessory subunit; DKFZp781H21101; HOKPP; MGC102685; MGC129924; Minimum potassium ion channel-related peptide 2; MinK-related peptide 2; MiRP2; Potassium channel subunit beta MiRP2; potassium channel, voltage gated subfamily E regulatory beta subunit 3; Potassium voltage-gated channel subfamily E member 3; potassium voltage-gated channel, Isk-related family, member 3; voltage-gated K+ channel subunit MIRP2
Gene Aliases: HOKPP; HYPP; KCNE3; MiRP2
UniProt ID: (Human) Q9Y6H6
Entrez Gene ID: (Human) 10008
Molecular Function: ion channel potassium channel transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.