Invitrogen
Manufacturer Code:PA577607
Catalog # PIPA577607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)IVEDWQQKYRSQILHLEDSK corresponding to amino acid residues 88-107 of rat KCNE2 (MiRP1) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cardiac voltage-gated potassium channel accessory subunit 2; minimum potassium ion channel related peptide 1; Minimum potassium ion channel-related peptide 1; MinK-related peptide 1; minK-related peptide-1; MiRP1; Potassium channel subunit beta MiRP1; potassium channel subunit, MiRP1; potassium channel, voltage gated subfamily E regulatory beta subunit 2; Potassium voltage-gated channel subfamily E member 2; potassium voltage-gated channel, Isk-related family, member 2; potassium voltage-gated channel, Isk-related subfamily, gene 2; voltage-gated K+ channel subunit MIRP1
Gene Aliases: 2200002I16Rik; ATFB4; AW048273; KCNE2; LQT5; LQT6; MIRP1
UniProt ID: (Human) Q9Y6J6, (Rat) P63161, (Mouse) Q9D808
Entrez Gene ID: (Human) 9992, (Rat) 171138, (Mouse) 246133
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.