Invitrogen
Manufacturer Code:PA559946
Catalog # PIPA559946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KCNE1L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMME syndrome candidate gene 2 protein; AMMECR2 protein; cardiac voltage-gated potassium channel accessory subunit 5; KCNE1-like; KCNE5 AMME syndrome candidate gene 2 protein AMMECR2 protein cardiac voltage-gated potassium channel accessory subunit 5 potassium voltage-gated channel subfamily E member 1-like protein potassium voltage-gated channel Isk-related family member 1-like; Potassium channel subunit beta MiRP4; potassium channel, voltage gated subfamily E regulatory beta subunit 5; Potassium voltage-gated channel subfamily E member 1-like protein; Potassium voltage-gated channel subfamily E regulatory beta subunit 5; potassium voltage-gated channel, Isk-related family, member 1-like
Gene Aliases: AMMECR2; KCNE1L; KCNE5
UniProt ID: (Human) Q9UJ90
Entrez Gene ID: (Human) 23630
Molecular Function:
ion channel
potassium channel
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.