Invitrogen
Manufacturer Code:PA5114298
Catalog # PIPA5114298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal region of human KCNE1L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMME syndrome candidate gene 2 protein; AMME syndrome candidate gene 2 protein AMMECR2 AMMECR2 protein cardiac voltage-gated potassium channel accessory subunit 5 KCNE1L KCNE1-like KCNE5 Potassium channel subunit beta MiRP4 potassium channel voltage gated subfamily E regulatory beta subunit 5 potassium channel voltage-gated Isk-related subfamily E regulatory beta subunit 5 Potassium voltage-gated channel subfamily E member 1-like protein Potassium voltage-gated channel subfamily E regulatory beta subunit 5 potassium voltage-gated channel subfamily E regulatory subunit 5 potassium voltage-gated channel Isk-related family member 1-like voltage-gated potassium channel accessory subunit 5; AMMECR2 protein; cardiac voltage-gated potassium channel accessory subunit 5; KCNE1-like; Potassium channel subunit beta MiRP4; potassium channel, voltage gated subfamily E regulatory beta subunit 5; Potassium voltage-gated channel subfamily E member 1-like protein; Potassium voltage-gated channel subfamily E regulatory beta subunit 5; potassium voltage-gated channel, Isk-related family, member 1-like
Gene Aliases: AMMECR2; KCNE1L; KCNE5
UniProt ID: (Human) Q9UJ90
Entrez Gene ID: (Human) 23630
Molecular Function: ion channel potassium channel transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.