Abnova
Manufacturer Code:PAB23924
Catalog # 89-125-631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human KCNE1L. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant KCNE1L.
For Research Use Only
Protein Aliases: AMME syndrome candidate gene 2 protein; AMMECR2 protein; cardiac voltage-gated potassium channel accessory subunit 5; KCNE1-like; KCNE5; Potassium channel subunit beta MiRP4; potassium channel, voltage gated subfamily E regulatory beta subunit 5; Potassium voltage-gated channel subfamily E member 1-like protein; Potassium voltage-gated channel subfamily E regulatory beta subunit 5; potassium voltage-gated channel, Isk-related family, member 1-like
Gene Aliases: AMMECR2; KCNE1L; KCNE5
UniProt ID: (Human) Q9UJ90
Entrez Gene ID: (Human) 23630
Molecular Function:
ion channel
potassium channel
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.