Proteintech
Manufacturer Code:138671AP150UL
Catalog # 50-172-9053
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KCNE1L Fusion Protein Ag4849 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMME syndrome candidate gene 2 protein; AMMECR2 KCNE1 like KCNE1L KCNE5; AMMECR2 protein; cardiac voltage-gated potassium channel accessory subunit 5; KCNE1-like; MinK-like protein; Potassium channel subunit beta MiRP4; potassium channel, voltage gated subfamily E regulatory beta subunit 5; Potassium voltage-gated channel subfamily E member 1-like protein; Potassium voltage-gated channel subfamily E regulatory beta subunit 5; potassium voltage-gated channel, Isk-related family, member 1-like; voltage-gated potassium channel accessory subunit 5
Gene Aliases: 1500015C14Rik; AMMECR2; KCNE1L; KCNE5; Mink
UniProt ID: (Human) Q9UJ90, (Mouse) Q9QZ26
Entrez Gene ID: (Human) 23630, (Mouse) 66240, (Rat) 681190
Molecular Function:
ion channel
potassium channel
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.