Proteintech
Manufacturer Code:151501AP150UL
Catalog # 50-173-0023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KCNE1 Fusion Protein Ag3495 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cardiac delayed rectifier potassium channel protein; Delayed rectifier potassium channel subunit IsK; IKs producing slow voltage-gated potassium channel subunit beta Mink; ISK JLNS JLNS2 KCNE1 LQT2/5 LQT5 Minimal potassium channel MinK; Minimal potassium channel; mink; mISK; potassium (K+) channel protein, slowly activating (Isk); potassium channel, voltage gated subfamily E regulatory beta subunit 1; potassium voltage-gated channel Shaw-related subfamily member 1; Potassium voltage-gated channel subfamily E member 1; potassium voltage-gated channel, Isk-related family, member 1; potassium voltage-gated channel, Isk-related subfamily, member 1; voltage gated potassiun channel accessory subunit
Gene Aliases: ISK; JLNS; JLNS2; KCNE1; LQT2/5; LQT5; m; MinK; nmf190
UniProt ID: (Human) P15382, (Mouse) P23299, (Rat) P15383
Entrez Gene ID: (Human) 3753, (Mouse) 16509, (Rat) 25471
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.