Novus Biologicals
Manufacturer Code:NBP2831020.1ML
Catalog # NB031031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human KCNB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Delayed rectifier potassium channel 1; Delayed rectifier potassium channel 1 delayed rectifier potassium channel Kv2.1 DRK1 h-DRK1 h-DRK1 K(+) channel Kv2.1 potassium channel protein DRK1 potassium voltage-gated channel subfamily B member 1 potassium voltage-gated channel Shab-related subfamily member 1 Voltage-gated potassium channel subunit Kv2.1; DRK1; Potassium voltage-gated channel subfamily B member 1; potassium voltage-gated channel, Shab-related subfamily, member 1; Voltage-gated potassium channel subunit Kv2.1
Gene Aliases: DRK1; KCNB1; Kv2.1
UniProt ID: (Human) Q14721
Entrez Gene ID: (Human) 3745
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.