Proteintech
Manufacturer Code:178901AP150UL
Catalog # 50-173-1857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KCNAB2 Fusion Protein Ag12096 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AKR6A5 HKvbeta2 HKvbeta2.1 HKvbeta2.2 K(+) channel subunit beta 2 KCNA2B KCNAB2 KCNK2 KV BETA 2 TREK TREK1; hKvbeta2; K(+) channel subunit beta-2; Kv-beta-2; Neuroimmune protein F5; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 2; potassium channel, voltage gated subfamily A regulatory beta subunit 2; potassium voltage gated channel, shaker related subfamily, beta member 2; potassium voltage-gated channel, shaker-related subfamily, beta member 2; Voltage-gated potassium channel subunit beta-2
Gene Aliases: AKR6A5; Ckbeta2; F5; HKvbeta2; HKvbeta2.1; HKvbeta2.2; I2rf5; KCNA2B; KCNAB2; Kcnb3; KCNK2; KV-BETA-2
UniProt ID: (Human) Q13303, (Mouse) P62482, (Rat) P62483
Entrez Gene ID: (Human) 8514, (Mouse) 16498, (Rat) 29738
Molecular Function:
ion channel
oxidoreductase
potassium channel
reductase
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.