Novus Biologicals
Manufacturer Code:NBP304425100UL
Catalog # NB095933
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human KCNA10 (NP_005540.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cyclic GMP gated potassium channel; cyclic GMP gated potassium channel Kcn1 Kv1.8 potassium voltage-gated channel subfamily A member 10 potassium voltage-gated channel shaker-related subfamily member 10 Voltage-gated potassium channel subunit Kv1.8; potassium channel, voltage gated shaker related subfamily A, member 10; Potassium voltage-gated channel subfamily A member 10; potassium voltage-gated channel, shaker-related subfamily, member 10; Voltage-gated potassium channel subunit Kv1.8
Gene Aliases: Kcn1; KCNA10; Kv1.8
UniProt ID: (Human) Q16322
Entrez Gene ID: (Human) 3744
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.