R&D Systems
Manufacturer Code:MAB9030
Catalog # MAB9030
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 891526 |
Immunogen | NS0 mouse myeloma cell line transfected with human SLC12A7 Met1-Ser1083 Accession # Q9Y666 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp434F076 KCC4Electroneutral potassium-chloride cotransporter 4 K-Cl cotransporter 4 potassium/chloride transporter KCC4 SLC12A7 solute carrier family 12 (potassium/chloride transporters) member 7 solute carrier family 12 member 7; Electroneutral potassium-chloride cotransporter 4; K-Cl cotransporter 4; potassium/chloride transporter KCC4; solute carrier family 12 (potassium/chloride transporter), member 7; solute carrier family 12 (potassium/chloride transporters), member 7; Solute carrier family 12 member 7
Gene Aliases: KCC4; SLC12A7
UniProt ID: (Human) Q9Y666
Entrez Gene ID: (Human) 10723
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.