Invitrogen
Manufacturer Code:PA559666
Catalog # PIPA559666
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KCC4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Electroneutral potassium-chloride cotransporter 4; furosemide-sensitive KCl cotransporter 4; K-Cl cotransporter 4; K-Cl cotransporter KCC4; KCC4 K-Cl cotransporter 4 electroneutral potassium-chloride cotransporter 4 potassium/chloride transporter KCC4 solute carrier family 12 member 7; potassium/chloride transporter KCC4; slc12a7 {ECO:0000312|EMBL:AAH86339.1, ECO:0000312|RGD:1359672}; solute carrier family 12 (potassium/chloride transporter), member 7; solute carrier family 12 (potassium/chloride transporters), member 7; Solute carrier family 12 member 7
Gene Aliases: AA408796; D13Ertd261e; KCC4; SLC12A7
UniProt ID: (Human) Q9Y666, (Rat) Q5RK27, (Mouse) Q9WVL3
Entrez Gene ID: (Human) 10723, (Rat) 308069, (Mouse) 20499
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.