Invitrogen
Manufacturer Code:PA556975
Catalog # PIPA556975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KCC3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Electroneutral potassium-chloride cotransporter 3; K-Cl cotransporter 3; potassium chloride cotransporter 3; potassium chloride cotransporter KCC3a-S3; potassium-chloride transporter-3a; potassium-chloride transporter-3b; SLC12A6 KCC3 solute carrier family 12 (potassium/chloride transporters) member 6 ACCPN DKFZP434D2135 KCC3A KCC3B agenesis of corpus callosum and peripheral neuropathy (Andermann syndrome) potassium chloride cotransporter 3 potassium chloride c; solute carrier family 12 (potassium/chloride transporter), member 6; solute carrier family 12 (potassium/chloride transporters), member 6; Solute carrier family 12 member 6
Gene Aliases: ACCPN; KCC3; KCC3A; KCC3B; SLC12A6
UniProt ID: (Human) Q9UHW9
Entrez Gene ID: (Human) 9990
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.