Proteintech
Manufacturer Code:195651AP150UL
Catalog # 50-173-2458
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; Furosemide-sensitive K-Cl cotransporter; hKCC2; hKCC2 K Cl cotransporter 2 KCC2 KIAA1176 Neuronal K Cl cotransporter SLC12A5; K-Cl cotransporter 2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; mKCC2; Neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; solute carrier family 12 (potassium/chloride transporter), member 5; Solute carrier family 12 member 5; solute carrier family 12, member 5
Gene Aliases: EIEE34; EIG14; hKCC2; KCC2; KIAA1176; mKIAA1176; SLC12A5
UniProt ID: (Human) Q9H2X9, (Mouse) Q91V14, (Rat) Q63633
Entrez Gene ID: (Human) 57468, (Mouse) 57138, (Rat) 171373
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.