Invitrogen
Manufacturer Code:PA577471
Catalog # PIPA577471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)HAPDNFRELVHIK corresponding to amino acid residues 998-1010 of rat KCC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Electroneutral potassium-chloride cotransporter 1; Erythroid K-Cl cotransporter 1; Furosemide-sensitive K-Cl cotransporter; hKCC1; K-Cl Co-transporter-1; K-Cl cotransporter KCC1; KCC1; mKCC1; potassium/chloride cotransporter 1; rKCC1; solute carrier family 12 (potassium/chloride transporter), member 4; solute carrier family 12 (potassium/chloride transporters), member 4; Solute carrier family 12 member 4; solute carrier family 12, member 4
Gene Aliases: AW546649; CTC-479C5.17; hKCC1; KCC1; RBCKCC1; SLC12A4
UniProt ID: (Human) F5H066, (Mouse) Q9JIS8, (Rat) Q63632
Entrez Gene ID: (Human) 6560, (Mouse) 20498, (Rat) 29501
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.