Abnova
Manufacturer Code:PAB21700
Catalog # 89-123-399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human KBTBD5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant KBTBD5.
For Research Use Only
Protein Aliases: FLJ32015; kelch repeat and BTB (POZ) domain containing 5; Kelch repeat and BTB domain-containing protein 5; kelch-like family member 40; Kelch-like protein 40; MGC125350; nemaline myopathy type 8; Sarcosynapsin; SRYP
Gene Aliases: KBTBD5; KLHL40; NEM8; SRYP; SYRP
UniProt ID: (Human) Q2TBA0
Entrez Gene ID: (Human) 131377
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.