Invitrogen
Manufacturer Code:PA571638
Catalog # PIPA571638
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 409-438 amino acids from the C-terminal region of human hCG&lowbar1645727 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: kelch repeat and BTB (POZ) domain containing 13; Kelch repeat and BTB domain-containing protein 13; Kelch repeat and BTB domain-containing protein 13 KBTBD13; nemaline myopathy type 6
Gene Aliases: HCG1645727; KBTBD13; NEM6
UniProt ID: (Human) C9JR72
Entrez Gene ID: (Human) 390594
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.