Invitrogen
Manufacturer Code:PA570562
Catalog # PIPA570562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of mouse KBP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KBP TTC20 KIF1-binding protein; KIF-binding protein; KIF1 binding protein; KIF1-binding protein; KIF1-binding protein-like protein; kinesin binding protein; Kinesin family binding protein
Gene Aliases: 0710007C18Rik; 2510003E04Rik; KBP; KIAA1279; KIF1BP; KIFBP; mKIAA1279; TTC20
UniProt ID: (Human) Q96EK5, (Bovine) Q3SYS9, (Mouse) Q6ZPU9, (Rat) Q4G074
Entrez Gene ID: (Human) 26128, (Rabbit) 100346136, (Bovine) 527433, (Horse) 100063227, (Dog) 479232, (Mouse) 72320, (Guinea pig) 100720078, (Rat) 606294
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.