OriGene
Manufacturer Code:CF503814
Catalog # 50-166-2403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5E1 |
Immunogen | Full length human recombit protein of human KATNB1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: KAT LIS6; katanin (80 kDa); katanin p80 (WD repeat containing) subunit B 1; katanin p80 (WD40-containing) subunit B 1; Katanin p80 subunit B1; Katanin p80 WD40 repeat-containing subunit B1; katanin p80 WD40-containing subunit B1; KATNB1; p80 katanin
Gene Aliases: KAT; KATNB1; LIS6
UniProt ID: (Human) Q9BVA0
Entrez Gene ID: (Human) 10300
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.