Novus Biologicals
Manufacturer Code:NBP203555
Catalog # NBP203555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A7 |
Immunogen | Full length human recombinant protein of human KATNAL1 (NP_115492) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.6.4.3 katanin p60 ATPase-containing subunit A-like 1 katanin p60 subunit A-like 1MGC2599 p60 katanin-like 1; katanin catalytic subunit A like 1; Katanin p60 ATPase-containing subunit A-like 1; katanin p60 subunit A like 1; Katanin p60 subunit A-like 1; KATNAL1; p60 katanin-like 1
Gene Aliases: KATNAL1
UniProt ID: (Human) Q9BW62
Entrez Gene ID: (Human) 84056
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.