OriGene
Manufacturer Code:CF502344
Catalog # 50-166-1255
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A7 |
Immunogen | Full length human recombit protein of human KATNAL1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: katanin catalytic subunit A like 1; katanin catalytic subunit A1 like 1; Katanin p60 ATPase-containing subunit A-like 1; katanin p60 ATPase-containing subunit A-like 1 {ECO:0000255|HAMAP-Rule:MF_03024}; katanin p60 subunit A like 1; Katanin p60 subunit A-like 1; katanin p60 subunit A-like 1 {ECO:0000255|HAMAP-Rule:MF_03024}; KATNAL1; p60 katanin-like 1; p60 katanin-like 1 {ECO:0000255|HAMAP-Rule:MF_03024}
Gene Aliases: KATNAL1
UniProt ID: (Human) Q9BW62, (Rat) Q5XIK7
Entrez Gene ID: (Human) 84056, (Rat) 288449
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.