Invitrogen
Manufacturer Code:PA5102975
Catalog # PIPA5102975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KATNA1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HAMAP-Rule:MF_03023} katanin catalytic subunit A1 katanin p60 (ATPase containing) subunit A 1 katanin p60 (ATPase-containing) subunit A 1 katanin p60 (ATPase-containing) subunit A1 katanin p60 ATPase-containing subunit A1 katanin p60 ATPase-containing subunit A1 {ECO:0000255 katanin p60 subunit A 1 Katanin p60 subunit A1 katanin p60 subunit A1 {ECO:0000255 Katna1 lipotransin microtubule severing protein p60 katanin p60 katanin {ECO:0000255; katanin p60 (ATPase containing) subunit A 1; katanin p60 (ATPase-containing) subunit A 1; Katanin p60 ATPase-containing subunit A1; katanin p60 ATPase-containing subunit A1 {ECO:0000255|HAMAP-Rule:MF_03023}; katanin p60 subunit A 1; Katanin p60 subunit A1; katanin p60 subunit A1 {ECO:0000255|HAMAP-Rule:MF_03023}; KATNA1; lipotransin; microtubule severing protein; p60 katanin; p60 katanin {ECO:0000255|HAMAP-Rule:MF_03023}
Gene Aliases: KATNA1
UniProt ID: (Human) O75449, (Rat) Q6E0V2
Entrez Gene ID: (Human) 11104, (Rat) 292464, (Mouse) 23924
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.