Abnova
Manufacturer Code:PAB26944
Catalog # 89-128-590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human KAT2A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of KAT2A.
For Research Use Only
Protein Aliases: GCN5; GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2; GCN5L2; General control of amino acid synthesis protein 5-like 2; General control of amino acid synthesis, yeast, homolog-like 2; hGCN5; Histone acetyltransferase GCN5; Histone acetyltransferase KAT2A; Histone glutaryltransferase KAT2A; Histone succinyltransferase KAT2A; hsGCN5; K(lysine) acetyltransferase 2A; Lysine acetyltransferase 2A; MGC102791; PCAF-b; STAF97
Gene Aliases: GCN5; GCN5L2; hGCN5; KAT2A; PCAF-b
UniProt ID: (Human) Q92830
Entrez Gene ID: (Human) 2648
Molecular Function:
DNA binding protein
acetyltransferase
chromatin/chromatin-binding protein
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.