Invitrogen
Manufacturer Code:PA559524
Catalog # PIPA559524
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KARS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: lysine tRNA ligase; Lysine--tRNA ligase; LysRS; Lysyl-tRNA synthetase; Lysyl-tRNA synthetase EC 6.1.1.6 Lysine--tRNA ligase LysRS KARS KIAA0070
Gene Aliases: AA589550; AL024334; AL033315; AL033367; CMTRIB; D8Ertd698e; D8Wsu108e; DFNB89; KARS; KARS1; KARS2; KIAA0070; KRS; LysRS; mKIAA0070
UniProt ID: (Human) Q15046, (Rat) Q5XIM7, (Mouse) Q99MN1
Entrez Gene ID: (Human) 3735, (Rat) 292028, (Mouse) 85305
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.