Invitrogen
Manufacturer Code:MA535678
Catalog # PIMA535678
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1211 |
Immunogen | A synthesized peptide derived from human KIFAP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: dJ190I16.1 FLA3 KAP-1 KAP3 KAP-3 KIF3AP KIFAP3 Kinesin-associated protein 3 SMAP Smg GDS-associated protein Smg-GDS; KAP-3; Kinesin-associated protein 3; small G protein GDP dissociation stimulator; Smg GDS-associated protein
Gene Aliases: dJ190I16.1; FLA3; KAP-1; KAP-3; KAP3; KIF3AP; KIFAP3; SMAP; Smg-GDS
UniProt ID: (Human) Q92845, (Mouse) P70188
Entrez Gene ID: (Human) 22920, (Rat) 289168, (Mouse) 16579
Molecular Function:
cytoskeletal protein
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.