Invitrogen
Manufacturer Code:PA554878
Catalog # PIPA554878
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KAP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KAP-3; KAP-3 KAP3 SMAP Smg-GDS dJ190I16.1 small G protein GDP dissociation stimulator smg GDS-associated protein; Kinesin-associated protein 3; small G protein GDP dissociation stimulator; Smg GDS-associated protein
Gene Aliases: dJ190I16.1; FLA3; KAP-1; KAP-3; KAP3; KIF3AP; KIFAP3; SMAP; Smg-GDS
UniProt ID: (Human) Q92845, (Mouse) P70188
Entrez Gene ID: (Human) 22920, (Mouse) 16579
Molecular Function: cytoskeletal protein microtubule binding motor protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.