R&D Systems
Manufacturer Code:MAB7785
Catalog # MAB7785
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 821912 |
Immunogen | <EM>E. coli</EM>-derived recombinant human KAP1 Lys266-Ser624 Accession # Q13263 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E3 SUMO-protein ligase TRIM28; E3 SUMO-protein ligase TRIM28 EC 6.3.2.- FLJ29029 KAP-1 KAP1KRAB-associated protein 1 KRIP-1 Nuclear corepressor KAP-1 RNF96 RNF96KRAB-interacting protein 1 TF1B TIF1B TIF1-beta TIF1BRING finger protein 96 transcription intermediary factor 1-beta transcriptional intermediary factor 1-beta TRIM28 tripartite motif containing 28 tripartite motif-containing 28 Tripartite motif-containing protein 28; KAP-1; KRAB [Kruppel-associated box domain]-associated protein 1; KRAB-associated protein 1; KRAB-interacting protein 1; KRIP-1; Nuclear corepressor KAP-1; protein phosphatase 1, regulatory subunit 157; RING finger protein 96; RING-type E3 ubiquitin transferase TIF1-beta; TIF1-beta; Transcription intermediary factor 1-beta; transcriptional intermediary factor 1-beta; Tripartite motif-containing protein 28
Gene Aliases: KAP1; PPP1R157; RNF96; TF1B; TIF1B; TRIM28
UniProt ID: (Human) Q13263
Entrez Gene ID: (Human) 10155
Molecular Function: transcription cofactor ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.