Invitrogen
Manufacturer Code:PA542085
Catalog # PIPA542085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ABP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABP; amiloride binding protein 1 (amine oxidase (copper-containing)); Amiloride binding protein 1 (amine oxidase (copper-containing)) ABP AOC1 DAO DAO1 KAO ABP1; amiloride binding protein 1 (amine oxidase, copper-containing); amiloride binding protein/diamine oxidase; Amiloride-binding protein 1; Amiloride-sensitive amine oxidase; Amiloride-sensitive amine oxidase [copper-containing]; Amine oxidase copper domain-containing protein 1; DAO; Diamine oxidase; Histaminase; KAO; Kidney amine oxidase
Gene Aliases: 1600012D06Rik; ABP; ABP1; AOC1; DAO; DAO1; KAO; si:ch211-286c5.2; zgc:154101
UniProt ID: (Pig) Q9TRC7, (Human) P19801
Entrez Gene ID: (Horse) 100063930, (Pig) 100517436, (Human) 26, (Bovine) 509751, (Rabbit) 100347103, (Dog) 475536, (Mouse) 76507, (Rat) 65029, (Guinea pig) 100735646, (Zebrafish) 555401
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.