Invitrogen
Manufacturer Code:PA552356
Catalog # PIPA552356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KANSL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: centromere protein 36; hMSL1v1; KAT8 regulatory NSL complex subunit 1; male-specific lethal 1 homolog; MLL1/MLL complex subunit KANSL1; MSL1 homolog 1; Non-specific lethal 1 homolog; NSL complex protein NSL1
Gene Aliases: CENP-36; hMSL1v1; KANSL1; KDVS; KIAA1267; MSL1V1; NSL1
UniProt ID: (Human) Q7Z3B3
Entrez Gene ID: (Human) 284058
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.