Novus Biologicals
Manufacturer Code:NBP15636620UL
Catalog # NBP15636620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ANKRD47 The peptide sequence was selected from the N terminal of ANKRD47. Peptide sequence GPAQLQLVREQMAAALRRLRELEDQARTLPELQEQVRALRAEKARLLAGR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANKRD47 ankyrin repeat domain 47 Ankyrin repeat domain-containing protein 47 FLJ46061 kidney ankyrin repeat-containing protein 3 KN motif and ankyrin repeat domain-containing protein 3 KN motif and ankyrin repeat domains 3; ankyrin repeat domain 47; Ankyrin repeat domain-containing protein 47; kidney ankyrin repeat-containing protein 3; KN motif and ankyrin repeat domain-containing protein 3
Gene Aliases: ANKRD47; KANK3
UniProt ID: (Human) Q6NY19
Entrez Gene ID: (Human) 256949
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.