Abnova
Manufacturer Code:PAB20832
Catalog # 89-122-330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human KANK2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant KANK2.
For Research Use Only
Protein Aliases: ANKRD25; Ankyrin repeat domain-containing protein 25; DKFZp434N161; FLJ20004; KIAA1518; kidney ankyrin repeat-containing protein 2; KN motif and ankyrin repeat domain-containing protein 2; Matrix-remodeling-associated protein 3; MGC119707; MXRA3; SIP; SRC-1-interacting protein; SRC-interacting protein; SRC1-interacting protein
Gene Aliases: ANKRD25; KANK2; KIAA1518; MXRA3; PPKWH; SIP
UniProt ID: (Human) Q63ZY3
Entrez Gene ID: (Human) 25959
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.