Proteintech
Manufacturer Code:205461AP150UL
Catalog # 50-173-2951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANKRD25 KANK2 KIAA1518 MXRA3 SIP SRC 1 interacting protein SRC1 interacting protein; ankyrin repeat domain 25; Ankyrin repeat domain-containing protein 25; kidney ankyrin repeat-containing protein 2; KN motif and ankyrin repeat domain-containing protein 2; Matrix-remodeling-associated protein 3; SIP; SRC-1-interacting protein; SRC-interacting protein; SRC1-interacting protein
Gene Aliases: AI504612; ANKRD25; BC010245; KANK2; KIAA1518; MXRA3; PPKWH; RGD1563532; SIP
UniProt ID: (Human) Q63ZY3, (Rat) D3ZD05, (Mouse) Q8BX02
Entrez Gene ID: (Human) 25959, (Rat) 100361376, (Mouse) 235041
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.