Invitrogen
Manufacturer Code:PA565999
Catalog # PIPA565999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human KANK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ankyrin repeat domain-containing protein 15; KANK1 KN motif and ankyrin repeat domains 1 Kidney ankyrin repeat-containing protein KANK Ankyrin repeat domain-containing protein 15 ANKRD15; Kidney ankyrin repeat-containing protein; KN motif and ankyrin repeat domain-containing protein 1
Gene Aliases: ANKRD15; CPSQ2; KANK; KANK1; KIAA0172
UniProt ID: (Human) Q14678
Entrez Gene ID: (Human) 23189
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.