Novus Biologicals
Manufacturer Code:NBP176852
Catalog # NBP176852
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of the human Grik4. The immunogen is located within amino acids 30 - 80 of Grik4. Amino Acid Squence: RINRAPERLGKAK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAA1; EAA1 Excitatory amino acid receptor 1 glutamate receptor KA1 Glutamate receptor KA-1 glutamate receptor ionotropic kainate 4 glutamate receptor ionotropic kainate 4 KA1GRIK; Excitatory amino acid receptor 1; GluK4; Glutamate receptor ionotropic, kainate 4; Glutamate receptor KA-1; glutamate receptor KA1; glutamate receptor, ionotropic, kainate 4; KA1
Gene Aliases: EAA1; GluK4; GRIK; GRIK4; KA1
UniProt ID: (Human) Q16099
Entrez Gene ID: (Human) 2900
Molecular Function: ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.