Invitrogen
Manufacturer Code:PA587049
Catalog # PIPA587049
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human CDH6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cadherin 6, type 2, K-cadherin (fetal kidney); Cadherin-6; Cadherin-6 Cadherin 6 Cadherin6 Kidney cadherin K-cadherin; K-cadherin; Kidney cadherin
Gene Aliases: CAD6; cadherin-6; CDH6; K-cadherin; KCAD
UniProt ID: (Human) P55285, (Rat) P55280, (Mouse) P97326
Entrez Gene ID: (Human) 1004, (Rat) 25409, (Mouse) 12563
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.