Invitrogen
Manufacturer Code:PA547373
Catalog # PIPA547373
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit human Cadherin-6 Thr22-Ala615 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: CAD6 KCAD cadherin-6; cadherin 6, type 2, K-cadherin (fetal kidney); Cadherin-6; K-cadherin; Kidney cadherin
Gene Aliases: CAD6; CDH6; KCAD
UniProt ID: (Human) P55285
Entrez Gene ID: (Human) 1004
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.