R&D Systems
Manufacturer Code:MAB5526
Catalog # MAB5526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 512709 |
Immunogen | <EM>E. coli</EM>-derived recombinant human JunD Lys47-Gly156 Accession # CAA40010 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: activator protein 1; activator protein 1 AP-1 jun D proto-oncogene JunD-FL isoform transcription factor jun-D; jun D proto-oncogene; JunD-FL isoform; Transcription factor jun-D
Gene Aliases: AP-1; JUND
UniProt ID: (Human) P17535
Entrez Gene ID: (Human) 3727
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.