Invitrogen
Manufacturer Code:PA517266
Catalog # PIPA517266
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues surrounding Pro169 of human JunB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activator protein 1; activator protein 1 transcription factor jun-B; jun B proto-oncogene; Jun-B oncogene; MyD21; Transcription factor jun-B
Gene Aliases: AP-1; Jun-b; JUNB
UniProt ID: (Human) P17275, (Rat) P24898, (Mouse) P09450
Entrez Gene ID: (Human) 3726, (Rat) 24517, (Mouse) 16477
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.