Invitrogen
Manufacturer Code:PA531846
Catalog # PIPA531846
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 231 of Human AHI1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abelson helper integration site 1 protein homolog; AHI-1; AHI-1 JBTS3 ORF1 dJ71N10.1 abelson helper integration site 1 protein homolog contatins SH3 and WD40 domains jouberin; contatins SH3 and WD40 domains; Jouberin
Gene Aliases: AHI-1; AHI1; dJ71N10.1; JBTS3; ORF1
UniProt ID: (Human) Q9H0H2
Entrez Gene ID: (Human) 54806
Molecular Function: RNA binding protein enzyme modulator esterase hydrolase kinase inhibitor kinase modulator mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.