R&D Systems
Manufacturer Code:MAB46991SP
Catalog # MAB46991SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 452524 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Jak3 Gly46-Thr209 Accession # P52333 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 JAK3_HUMAN JAK-3EC 2.7.10.2 JAKL JAKLtyrosine-protein kinase JAK3 Janus kinase 3Leukocyte janus kinase LJAK L-JAKJanus kinase 3 (a protein tyrosine kinase leukocyte); JAK-3; Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); L-JAK; Leukocyte janus kinase; Tyrosine-protein kinase JAK3
Gene Aliases: JAK-3; JAK3; JAK3_HUMAN; JAKL; L-JAK; LJAK
UniProt ID: (Human) P52333
Entrez Gene ID: (Human) 3718
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.