R&D Systems
Manufacturer Code:IC4260G100UG
Catalog # IC4260G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 413104 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Jak1 Pro32-Phe286 Accession # P23458 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Rat MonoclonalAntibody
For Research Use Only
Protein Aliases: EC 2.7.10 JAK-1 JAK1Atyrosine-protein kinase JAK1 JAK1B Janus kinase 1EC 2.7.10.2 JTK3; JAK-1; Janus kinase 1; Tyrosine-protein kinase JAK1
Gene Aliases: JAK1; JAK1A; JAK1B; JTK3
UniProt ID: (Human) P23458
Entrez Gene ID: (Human) 3716
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.