R&D Systems
Manufacturer Code:AF602SP
Catalog # AF602SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <em>E. coli</em>-derived recombinant mouse Jak1 Ala14-Tyr55 (predicted) Accession # P52332 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 JAK-1 JAK1Atyrosine-protein kinase JAK1 JAK1B Janus kinase 1EC 2.7.10.2 JTK3; JAK-1; Janus kinase 1; Tyrosine-protein kinase JAK1
Gene Aliases: JAK1; JAK1A; JAK1B; JTK3
UniProt ID: (Human) P23458
Entrez Gene ID: (Human) 3716
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.