Novus Biologicals
Manufacturer Code:NB1001105
Catalog # NB1001105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-NRLTDYISKVKE corresponding to C-Terminus according to NP_006398.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor GTPase 6 interacting protein 5; ADP-ribosylation factor like GTPase 6 interacting protein 5; ADP-ribosylation factor-like 6 interacting protein 5; ADP-ribosylation factor-like protein 6-interacting protein 5; ADP-ribosylation factor-like protein 6-interacting protein 5 ADP-ribosylation-like factor 6 interacting protein 5 Aip-5 ARL-6-interacting protein 5 Cytoskeleton-related vitamin A-responsive protein Dermal papilla-derived protein 11 DERP11addicsin Glutamate transporter EAAC1-interacting protein glutamate transporter EEAC1-associated protein GTRAP3-18aip-5 hp22 HSPC127 JM5 jmx JWAcytoskeleton related vitamin A responsive protein PRA1 domain family 3 PRA1 family protein 3 PRA2 PRAF3dermal papilla derived protein 11 Prenylated Rab acceptor protein 2 Protein JWa putative MAPK activating protein PM27 Putative MAPK-activating protein PM27; ADP-ribosylation-like factor 6 interacting protein 5; aip-5; ARL-6-interacting protein 5; cytoskeleton related vitamin A responsive protein; Cytoskeleton-related vitamin A-responsive protein; dermal papilla derived protein 11; Dermal papilla-derived protein 11; Glutamate transporter EAAC1-interacting protein; glutamate transporter EEAC1-associated protein; GTRAP3-18; JM5; PRA1 domain family 3; PRA1 family protein 3; Prenylated Rab acceptor protein 2; Protein JWa; putative MAPK activating protein PM27; Putative MAPK-activating protein PM27
Gene Aliases: addicsin; ARL6IP5; DERP11; GTRAP3-18; hp22; HSPC127; jmx; JWA; PRA2; PRAF3; Yip6b
UniProt ID: (Human) O75915
Entrez Gene ID: (Human) 10550
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.