Invitrogen
Manufacturer Code:PA553923
Catalog # PIPA553923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human JTV-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aminoacyl tRNA synthase complex-interacting multifunctional protein 2; ARS interacting multifunctional protein 2; ARS-interacting multi-functional protein 2; JTV-1 JTV1 P38 ARS-interacting multi-functional protein 2 JTV1 aminoacyl tRNA synthase complex-interacting multifunctional protein 2 multisynthase complex auxiliary component p38 multisynthetase complex auxiliary component p38 protein JTV-1; JTV1; Multisynthase complex auxiliary component p38; multisynthetase complex auxiliary component p38; Protein JTV-1
Gene Aliases: AA407136; AA407225; AIMP2; Aimp2(p38); JTV-1; JTV1; P38; PRO0992
UniProt ID: (Human) Q13155, (Rat) Q32PX2, (Mouse) Q8R010
Entrez Gene ID: (Human) 7965, (Rat) 288480, (Mouse) 231872
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.