Novus Biologicals
Manufacturer Code:NBP304953100UL
Catalog # NB095192
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 31-105 of human JTB (NP_006685.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hJT HJTB HSPC222 jumping translocation breakpoint Jumping translocation breakpoint protein PAR PAR protein Prostate androgen-regulated protein protein JTB; Jumping translocation breakpoint protein; PAR protein; Prostate androgen-regulated protein; Protein JTB
Gene Aliases: hJT; HJTB; HSPC222; JTB; PAR
UniProt ID: (Human) O76095
Entrez Gene ID: (Human) 10899
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.